siRNA Details
Gene Symbol: Pctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017225.2 | NP_058921.1 | 1 | 114 | ||
XM_006247125.2 | XP_006247187.1 | 1 | 182 | |||
GenBank mRNA | AF040261.1 | - | 1 | 58 | ||
BC078854.1 | - | 1 | 119 | 7113616 |
