siRNA Details
| Gene Symbol: Psma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017278.1 | NP_058974.1 | 3 | 178 | ||
| GenBank mRNA | BC062233.1 | - | 3 | 175 | 6919336 | |
| D90265.1 | - | 3 | 178 | |||
| FQ212620.1 | - | 3 | 147 | |||
| FQ213606.1 | - | 3 | 168 | |||
| FQ215179.1 | - | 3 | 157 | |||
| FQ215378.1 | - | 3 | 161 | |||
| FQ216107.1 | - | 3 | 171 | |||
| FQ218243.1 | - | 210 | ||||
| FQ220925.1 | - | 3 | 143 | |||
| FQ222787.1 | - | 3 | 171 | |||
| FQ223714.1 | - | 3 | 147 | |||
| FQ225726.1 | - | 3 | 129 | |||
| M29859.1 | - | 3 | 178 | |||

Psma1
Genomic Map