siRNA Details
| Gene Symbol: FOXJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018416.2 | NP_060886.1 | 6 | 1840 | ||
| XM_011520760.2 | XP_011519062.1 | 6 | 1923 | |||
| XM_011520761.2 | XP_011519063.1 | 7 | 2024 | |||
| XM_017019640.1 | XP_016875129.1 | 5 | 587 | |||
| GenBank mRNA | AF155132.1 | - | 6 | 1244 | ||
| AF155133.1 | - | 6 | 1244 | |||
| BC126396.1 | - | 6 | 810 | 8992112 | ||
| BC136305.1 | - | 6 | 810 | 9020292 | ||

FOXJ2
Genomic Map