siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135562.1 | NP_001129034.1 | 2 | 148 | ||
| NM_021262.2 | NP_067085.1 | 2 | 115 | |||
| XM_006239965.2 | XP_006240027.1 | 2 | 144 | |||
| XM_008764572.1 | XP_008762794.1 | 2 | 148 | |||
| GenBank mRNA | AF171072.1 | - | 2 | 78 | ||
| BC062229.1 | - | 2 | 115 | 6918194 | ||
| BC158763.1 | - | 2 | 148 | 7109165 | ||
| BC161990.1 | - | 2 | 94 | 9032684 | ||
| FQ213063.1 | - | 2 | 95 | |||
| FQ215147.1 | - | 2 | 86 | |||

Acp1
Genomic Map