siRNA Details
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030499.1 | - | 4, 5 | 521 | ||
AB030500.1 | - | 4, 5 | 521 | |||
AB030501.1 | - | 4, 5 | 521 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030499.1 | - | 4, 5 | 521 | ||
AB030500.1 | - | 4, 5 | 521 | |||
AB030501.1 | - | 4, 5 | 521 |