siRNA Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022256.2 | NP_071592.1 | 150 | |||
| GenBank mRNA | AB028862.1 | - | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btc| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022256.2 | NP_071592.1 | 150 | |||
| GenBank mRNA | AB028862.1 | - | 150 | |||