siRNA Details
| Gene Symbol: Birc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022274.1 | NP_071610.1 | 2 | 159 | ||
| GenBank mRNA | AF276775.1 | - | 2 | 159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Birc5| Gene Symbol: Birc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022274.1 | NP_071610.1 | 2 | 159 | ||
| GenBank mRNA | AF276775.1 | - | 2 | 159 | ||