siRNA Details
| Gene Symbol: C5AR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271749.1 | NP_001258678.1 | 2 | 1387 | ||
| NM_001271750.1 | NP_001258679.1 | 2 | 1310 | |||
| NM_018485.2 | NP_060955.1 | 2 | 1349 | |||
| XM_011526736.2 | XP_011525038.1 | 3 | 2024 | |||
| GenBank mRNA | AB038237.1 | - | 2 | 1240 | ||
More Information
Set Membership: |
|

C5AR2
Genomic Map