siRNA Details
| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031120.2 | NP_112382.2 | 176 | |||
| GenBank mRNA | AY724496.1 | - | 184 | |||
| BC062015.1 | - | 176 | 5621495 | |||
| Z14030.1 | - | 184 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssr3| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031120.2 | NP_112382.2 | 176 | |||
| GenBank mRNA | AY724496.1 | - | 184 | |||
| BC062015.1 | - | 176 | 5621495 | |||
| Z14030.1 | - | 184 | ||||