siRNA Details
| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031550.1 | NP_113738.1 | 3 | 546 | ||
| GenBank mRNA | AF474974.1 | - | 587 | |||
| AF474975.1 | - | 587 | ||||
| AF474976.1 | - | 3 | 522 | |||
| AF474977.1 | - | 3 | 513 | |||
| AY679727.1 | - | 587 | ||||
| AY679728.1 | - | 3 | 522 | |||
| L81167.1 | - | 3 | 546 | |||

Cdkn2a
Genomic Map