siRNA Details
| Gene Symbol: Sh2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031621.1 | NP_113809.1 | 1 | 368 | ||
| GenBank mRNA | JX215366.1 | - | 1189 | |||
| U24652.1 | - | 3 | 215 | |||
| U24653.1 | - | 3 | 215 | |||
| U24654.1 | - | 3 | 215 | |||
| U24655.1 | - | 1 | 368 | |||

Sh2b3
Genomic Map