siRNA Details
Gene Symbol: Kap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052802.1 | NP_434689.1 | 2 | 250 | ||
XM_006237483.2 | XP_006237545.1 | 2 | 178 | |||
GenBank mRNA | U25808.1 | - | 2 | 250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052802.1 | NP_434689.1 | 2 | 250 | ||
XM_006237483.2 | XP_006237545.1 | 2 | 178 | |||
GenBank mRNA | U25808.1 | - | 2 | 250 |