siRNA Details
Gene Symbol: Nrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053346.1 | NP_445798.1 | 1 | 286 | ||
GenBank mRNA | BC087582.1 | - | 1 | 223 | 7315687 | |
U88958.1 | - | 1 | 286 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053346.1 | NP_445798.1 | 1 | 286 | ||
GenBank mRNA | BC087582.1 | - | 1 | 223 | 7315687 | |
U88958.1 | - | 1 | 286 |