siRNA Details
| Gene Symbol: Ran | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053439.1 | NP_445891.1 | 3 | 182 | ||
| XM_006249285.2 | XP_006249347.1 | 3 | 169 | |||
| GenBank mRNA | AF306457.1 | - | 3 | 182 | ||
| BC059123.1 | - | 3 | 197 | 6920235 | ||
| FQ220083.1 | - | 3 | 165 | |||
| FQ220326.1 | - | 3 | 165 | |||
| FQ220799.1 | - | 3 | 210 | |||
| FQ229342.1 | - | 3 | 134 | |||
| FQ229588.1 | - | 3 | 188 | |||
| FQ229761.1 | - | 3 | 173 | |||
| FQ231051.1 | - | 2 | 177 | |||
| FQ233248.1 | - | 3 | 136 | |||

Ran
Genomic Map