siRNA Details
| Gene Symbol: Nupr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053611.2 | NP_446063.1 | 1 | 172 | ||
| GenBank mRNA | AA685246.1 | - | 1 | 172 | ||
| AF014503.1 | - | 1 | 140 | |||
| FQ221101.1 | - | 1 | 134 | |||
| Gene Symbol: NEWGENE_69363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759924.1 | XP_008758146.1 | 1 | 223 | ||

Nupr1
Genomic Map