siRNA Details
| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053894.1 | NP_446346.1 | 1 | 373 | ||
| XM_008764734.1 | XP_008762956.1 | 2 | 943 | |||
| XM_008764735.1 | XP_008762957.1 | 2 | 270 | |||
| GenBank mRNA | U53449.1 | - | 1 | 373 | ||

Jdp2
Genomic Map