siRNA Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057137.1 | NP_476478.1 | 2 | 267 | ||
| XM_006256699.2 | XP_006256761.1 | 2 | 292 | |||
| GenBank mRNA | AF071501.1 | - | 2 | 267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ebp| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057137.1 | NP_476478.1 | 2 | 267 | ||
| XM_006256699.2 | XP_006256761.1 | 2 | 292 | |||
| GenBank mRNA | AF071501.1 | - | 2 | 267 | ||