siRNA Details
| Gene Symbol: Rab3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080580.2 | NP_542147.1 | 2 | 275 | ||
| XM_006242593.1 | XP_006242655.1 | 2 | 271 | |||
| XM_006242594.2 | XP_006242656.1 | 2 | 391 | |||
| GenBank mRNA | BC081741.1 | - | 2 | 275 | 7110109 | |
| M83681.1 | - | 1 | 103 | |||
| S68808.1 | - | 1 | 121 | |||
| U90206.1 | - | 1 | 166 | |||

Rab3d
Genomic Map