siRNA Details
| Gene Symbol: Fgf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130754.1 | NP_570110.1 | 1 | 78 | ||
| GenBank mRNA | AB078777.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf23| Gene Symbol: Fgf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130754.1 | NP_570110.1 | 1 | 78 | ||
| GenBank mRNA | AB078777.1 | - | 1 | 78 | ||