siRNA Details
| Gene Symbol: Cx3cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133534.1 | NP_598218.1 | 2 | 261 | ||
| XM_008766668.1 | XP_008764890.1 | 2 | 576 | |||
| GenBank mRNA | U04808.1 | - | 2 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cx3cr1| Gene Symbol: Cx3cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133534.1 | NP_598218.1 | 2 | 261 | ||
| XM_008766668.1 | XP_008764890.1 | 2 | 576 | |||
| GenBank mRNA | U04808.1 | - | 2 | 261 | ||