siRNA Details
| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138513.1 | NP_612522.1 | 2 | 141 | ||
| XM_006230075.2 | XP_006230137.1 | 4 | 615 | |||
| XM_008759674.1 | XP_008757896.1 | 5 | 470 | |||
| GenBank mRNA | M11596.1 | - | 2 | 141 | ||

Calcb
Genomic Map