siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138845.2 | NP_620200.2 | 23, 24 | 5252 | ||
| XM_006242061.2 | XP_006242123.1 | 20, 21 | 5532 | |||
| XM_006242063.2 | XP_006242125.1 | 19, 20 | 5360 | |||
| XM_006242064.2 | XP_006242126.1 | 18, 19 | 5201 | |||
| GenBank mRNA | AF275151.1 | - | 15, 16 | 2007 | ||
| BC061751.1 | - | 7, 8 | 928 | |||

Tnrc6b
Genomic Map