siRNA Details
| Gene Symbol: GPR173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018969.5 | NP_061842.1 | 2 | 776 | ||
| XM_011530798.2 | XP_011529100.1 | 2 | 793 | |||
| GenBank mRNA | AB040801.1 | - | 1 | 518 | ||
| BC009861.2 | - | 2 | 776 | 3936037 | ||
| BI602566.1 | - | 1 | 568 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027543, AY255602, BC043021, BC096520.
Set Membership: |
|

GPR173
Genomic Map