siRNA Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139104.1 | NP_620804.1 | 6, 7 | 821 | ||
| XM_006233668.2 | XP_006233730.1 | 6, 7 | 924 | |||
| XM_006233669.2 | XP_006233731.1 | 5, 6 | 478 | |||
| GenBank mRNA | AF223678.1 | - | 6, 7 | 821 | ||
| BC078725.1 | - | 6, 7 | 765 | 7127696 | ||

Egfl7
Genomic Map