siRNA Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172033.2 | NP_742030.1 | 6 | 601 | ||
| XM_006229787.2 | XP_006229849.1 | 7 | 839 | |||
| XM_006229788.2 | XP_006229850.1 | 6 | 1000 | |||
| XM_006229789.2 | XP_006229851.1 | 7 | 910 | |||
| GenBank mRNA | AF081582.1 | - | 6 | 601 | ||
| AF118562.1 | - | 6 | 571 | |||

Plekhb1
Genomic Map