siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 7 | 763 | ||
| GenBank mRNA | AK131793.1 | - | 7 | 565 | ||
| AK146444.1 | - | 7 | 568 | |||
| AK206711.1 | - | 1 | 37 | |||
| BC013498.1 | - | 7 | 548 | 4222450 | ||
| U07159.1 | - | 7 | 552 | |||

Acadm
Genomic Map