siRNA Details
| Gene Symbol: TAS2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019599.2 | NP_062545.1 | 1 | 1094 | ||
| GenBank mRNA | BC095521.1 | - | 1 | 1094 | 30706582 | |
| BC101729.1 | - | 1 | 848 | 8069235 | ||
| BC101731.1 | - | 1 | 848 | 8069237 | ||

TAS2R1
Genomic Map