siRNA Details
| Gene Symbol: MC3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019888.3 | NP_063941.3 | 1 | 545 | ||
| GenBank mRNA | AY227893.1 | - | 1 | 544 | ||
| BC069105.1 | - | 1 | 545 | 7216921 | ||
| BC069599.1 | - | 1 | 544 | 7262270 | ||
| BC096702.1 | - | 1 | 544 | 40016185 | ||
| BC096737.1 | - | 1 | 544 | 40016189 | ||
| BC098169.1 | - | 1 | 544 | 40016186 | ||
| BC098351.1 | - | 1 | 544 | 40016184 | ||
More Information
Set Membership: |
|

MC3R
Genomic Map