siRNA Details
Gene Symbol: Alk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007439.2 | NP_031465.2 | 1 | 1059 | ||
GenBank mRNA | AK051358.1 | - | 1 | 1060 | ||
AK158763.1 | - | 1 | 1381 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007439.2 | NP_031465.2 | 1 | 1059 | ||
GenBank mRNA | AK051358.1 | - | 1 | 1060 | ||
AK158763.1 | - | 1 | 1381 |