siRNA Details
| Gene Symbol: Ambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007443.3 | NP_031469.1 | 1 | 182 | ||
| GenBank mRNA | AK004907.1 | - | 1 | 183 | ||
| BC021660.1 | - | 1 | 201 | 4193922 | ||
| D28812.1 | - | 1 | 183 | |||
| X68680.1 | - | 1 | 144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ambp| Gene Symbol: Ambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007443.3 | NP_031469.1 | 1 | 182 | ||
| GenBank mRNA | AK004907.1 | - | 1 | 183 | ||
| BC021660.1 | - | 1 | 201 | 4193922 | ||
| D28812.1 | - | 1 | 183 | |||
| X68680.1 | - | 1 | 144 | |||