siRNA Details
| Gene Symbol: Aqp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109045.1 | NP_001102515.1 | 4 | 560 | ||
| NM_007474.2 | NP_031500.1 | 4 | 563 | |||
| GenBank mRNA | AF018952.1 | - | 4 | 545 | ||
| AK146660.1 | - | 3 | 242 | |||
| BC010982.1 | - | 4 | 505 | 4224850 | ||
| BY713653.1 | - | 4 | 565 | |||
| CF617211.1 | - | 4 | 466 | |||

Aqp8
Genomic Map