siRNA Details
Gene Symbol: Arf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007479.3 | NP_031505.1 | 3 | 467 | ||
GenBank mRNA | AK002302.1 | - | 3 | 490 | ||
AK013892.1 | - | 3 | 522 | |||
AK081686.1 | - | 3 | 467 | |||
AK153011.1 | - | 3 | 451 | |||
AK168793.1 | - | 3 | 488 | |||
AK208695.1 | - | 1 | 86 | |||
BC115650.1 | - | 3 | 346 | 40103144 | ||
BC115651.1 | - | 3 | 346 | 40103145 | ||
D87901.1 | - | 3 | 284 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
