siRNA Details
| Gene Symbol: Abca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007379.2 | NP_031405.2 | 4 | 416 | ||
| GenBank mRNA | AK169438.1 | - | 4 | 416 | ||
| X75927.2 | - | 4 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca2| Gene Symbol: Abca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007379.2 | NP_031405.2 | 4 | 416 | ||
| GenBank mRNA | AK169438.1 | - | 4 | 416 | ||
| X75927.2 | - | 4 | 320 | |||