siRNA Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007399.3 | NP_031425.2 | 2 | 380 | ||
| GenBank mRNA | AF011379.1 | - | 2 | 201 | ||
| BC057346.1 | - | 2 | 342 | |||
| BC066207.1 | - | 2 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam10| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007399.3 | NP_031425.2 | 2 | 380 | ||
| GenBank mRNA | AF011379.1 | - | 2 | 201 | ||
| BC057346.1 | - | 2 | 342 | |||
| BC066207.1 | - | 2 | 385 | |||