siRNA Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288578.1 | NP_001275507.1 | 3 | 284 | ||
| NM_007410.3 | NP_031436.2 | 3 | 302 | |||
| GenBank mRNA | AK002790.1 | - | 3 | 224 | ||
| AK076507.1 | - | 3 | 231 | |||
| AK146949.1 | - | 3 | 246 | |||
| AK159803.1 | - | 3 | 282 | |||
| AK206859.1 | - | 55 | ||||
| BC062879.1 | - | 3 | 228 | 5691327 | ||
| BC090978.1 | - | 3 | 225 | 30313517 | ||
| BY125542.1 | - | 3 | 302 | |||
| CF172597.1 | - | 3 | 284 | |||
| M84147.1 | - | 3 | 252 | |||

Adh5
Genomic Map