siRNA Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007422.3 | NP_031448.2 | 3, 4 | 471 | ||
| GenBank mRNA | AK004877.1 | - | 3, 4 | 424 | ||
| AK010263.1 | - | 3, 4 | 446 | |||
| AK028060.1 | - | 3, 4 | 471 | |||
| AK043273.1 | - | 3, 4 | 464 | |||
| AK148420.1 | - | 3, 4 | 453 | |||
| AK170279.1 | - | 3, 4 | 457 | |||
| BC139404.1 | - | 3, 4 | 383 | 8862426 | ||
| BC139417.1 | - | 3, 4 | 383 | 8862439 | ||
| L24554.1 | - | 3, 4 | 438 | |||

Adss
Genomic Map