siRNA Details
| Gene Symbol: Ap3d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007460.1 | NP_031486.1 | 6 | 716 | ||
| GenBank mRNA | AB004305.1 | - | 6 | 716 | ||
| AK141789.1 | - | 6 | 564 | |||
| AK155518.1 | - | 6 | 766 | |||
| BC022171.1 | - | 6 | 721 | |||
| BC037477.1 | - | 6 | 717 | |||
| BC048786.1 | - | 6 | 706 | 6489789 | ||
| BC053066.1 | - | 6 | 732 | 5704929 | ||

Ap3d1
Genomic Map