siRNA Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007399.3 | NP_031425.2 | 13 | 1920 | ||
| GenBank mRNA | AF011379.1 | - | 13 | 1741 | ||
| AK036599.1 | - | 7 | 964 | |||
| AK036883.1 | - | 5 | 608 | |||
| AK036918.1 | - | 5 | 609 | |||
| AK217253.1 | - | 2 | 141 | |||
| BC057346.1 | - | 13 | 1881 | |||

Adam10
Genomic Map