siRNA Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007405.2 | NP_031431.2 | 11, 12 | 2127 | ||
| GenBank mRNA | AK147516.1 | - | 11, 12 | 2127 | ||
| BC145101.1 | - | 10, 11 | 1960 | |||
| M93422.1 | - | 10, 11 | 2059 | |||
| M96653.1 | - | 10, 11 | 1949 | |||

Adcy6
Genomic Map