siRNA Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288578.1 | NP_001275507.1 | 5 | 479 | ||
| NM_007410.3 | NP_031436.2 | 5 | 497 | |||
| GenBank mRNA | AK002790.1 | - | 5 | 419 | ||
| AK076507.1 | - | 5 | 426 | |||
| AK146949.1 | - | 5 | 441 | |||
| BC062879.1 | - | 5 | 423 | 5691327 | ||
| BC090978.1 | - | 5 | 420 | 30313517 | ||
| CF172597.1 | - | 5 | 479 | |||
| M84147.1 | - | 5 | 447 | |||

Adh5
Genomic Map