siRNA Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007422.3 | NP_031448.2 | 7, 8 | 786 | ||
| GenBank mRNA | AK004877.1 | - | 7, 8 | 739 | ||
| AK010263.1 | - | 7, 8 | 761 | |||
| AK028060.1 | - | 7, 8 | 786 | |||
| AK148420.1 | - | 7, 8 | 768 | |||
| AK170279.1 | - | 7, 8 | 772 | |||
| BC139404.1 | - | 7, 8 | 698 | 8862426 | ||
| BC139417.1 | - | 7, 8 | 698 | 8862439 | ||
| L24554.1 | - | 7, 8 | 753 | |||

Adss
Genomic Map