siRNA Details
| Gene Symbol: Ambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007443.3 | NP_031469.1 | 9 | 1085 | ||
| GenBank mRNA | AK004907.1 | - | 9 | 1086 | ||
| BC021660.1 | - | 9 | 1104 | 4193922 | ||
| D28812.1 | - | 9 | 1086 | |||
| X68680.1 | - | 9 | 1047 | |||

Ambp
Genomic Map