siRNA Details
| Gene Symbol: Ap1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF068707.1 | - | 20 | 2272 | ||
| AK052384.1 | - | 19 | 2426 | |||
| AK147177.1 | - | 21 | 2311 | |||
| AK151844.1 | - | 21 | 2303 | |||
| BC058402.1 | - | 19 | 2317 | |||
| BC080798.1 | - | 20 | 2219 | |||
| BC108374.1 | - | 21 | 2298 | 4936738 | ||

Ap1g2
Genomic Map