siRNA Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077264.1 | NP_001070732.1 | 21 | 2750 | ||
| NM_007458.2 | NP_031484.1 | 22 | 2816 | |||
| GenBank mRNA | AK135424.1 | - | 21 | 2679 | ||
| AK155515.1 | - | 21 | 2687 | |||
| BC031433.1 | - | 22 | 2766 | 4481643 | ||

Ap2a1
Genomic Map