siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134480.1 | - | 10 | 1291 | ||
| AK137466.1 | - | 10 | 1320 | |||
| AK153934.1 | - | 10 | 1264 | |||
| AK160874.1 | - | 10 | 1288 | |||
| AK167315.1 | - | 10 | 1263 | |||
| AK167375.1 | - | 10 | 1304 | |||
| AK210073.1 | - | 4 | 385 | |||
| BC007133.1 | - | 10 | 1283 | 3594358 | ||
| U35846.1 | - | 10 | 1248 | |||

Api5
Genomic Map