siRNA Details
| Gene Symbol: Atm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007499.2 | NP_031525.2 | 3 | 252 | ||
| GenBank mRNA | AK139863.1 | - | 3 | 256 | ||
| BC138525.1 | - | 2 | 154 | 8861545 | ||
| BY725995.1 | - | 3 | 252 | |||
| U43678.1 | - | 2 | 154 | |||

Atm
Genomic Map