siRNA Details
| Gene Symbol: Atpif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF002718.1 | - | 2 | 194 | ||
| AK008346.1 | - | 2 | 192 | |||
| AK011886.1 | - | 2 | 191 | |||
| AK152389.1 | - | 3 | 1209 | |||
| AK161394.1 | - | 1 | 283 | |||
| AV462016.1 | - | 2 | 255 | |||
| BC012680.1 | - | 2 | 253 | 4217188 | ||
| BU055998.1 | - | 2 | 180 | |||
| CB848021.2 | - | 2 | 217 | |||

Atpif1
Genomic Map