siRNA Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044740.2 | NP_001038205.1 | 3, 4 | 747 | ||
| NM_007514.3 | NP_031540.2 | 3, 4 | 747 | |||
| GenBank mRNA | AK154621.1 | - | 3, 4 | 721 | ||
| AK155558.1 | - | 3, 4 | 722 | |||
| AK171369.1 | - | 3, 4 | 719 | |||
| BC021653.1 | - | 3, 4 | 742 | |||
| BC127082.1 | - | 3, 4 | 663 | 40111372 | ||
| CN710433.1 | - | 2, 3 | 354 | |||
| DQ086834.1 | - | 3, 4 | 702 | |||
| L03290.1 | - | 2, 3 | 549 | |||
| L11600.1 | - | 2, 3 | 549 | |||
| L29006.1 | - | 2, 3 | 1072 | |||
| M62838.1 | - | 3, 4 | 660 | |||

Slc7a2
Genomic Map