siRNA Details
| Gene Symbol: Nkx3-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007524.3 | NP_031550.2 | 1 | 485 | ||
| GenBank mRNA | BC021014.1 | - | 1 | 768 | ||
| BC145872.1 | - | 1 | 480 | 40131175 | ||
| BC145874.1 | - | 1 | 480 | 40131177 | ||
| U87957.1 | - | 1 | 485 | |||

Nkx3-2
Genomic Map