siRNA Details
| Gene Symbol: Bmp8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256019.1 | NP_001242948.1 | 4 | 1352 | ||
| NM_007558.3 | NP_031584.1 | 4 | 1352 | |||
| GenBank mRNA | AK157978.1 | - | 4 | 1014 | ||
| BC052168.1 | - | 4 | 1350 | 6334507 | ||
| M97017.1 | - | 4 | 900 | |||

Bmp8a
Genomic Map